APO455831145 Apollo Scientific Ltd

2,5-Dibromobenzoic acid

  • MDL number MFCD00016494
  • Cas Number : 610-71-9
  • Molecular Weight : 279.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-159
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/25-dibromobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/25-dibromobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart