APO455831148 Apollo Scientific Ltd

2,4-Dichloro-5-methoxyaniline

  • MDL number MFCD00974410
  • Cas Number : 98446-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-51
Synonym 5-Amino-2,4-dichloroanisole, 4,6-Dichloro-m-anisidine
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=30073

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=30073
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart