APO455831151 Apollo Scientific Ltd

3-Amino-1-bromoisoquinoline

  • MDL number MFCD00102193
  • Cas Number : 13130-79-5
  • Molecular Weight : 223.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 144-146
Iupac Name 1-bromo-3-isoquinolinamine
Inchi Code 1S/C9H7BrN2/c10-9-7-4-2-1-3-6(7)5-8(11)12-9/h1-5H,(H2,11,12)
Inchi Key PSQUIUNIVDKHJK-UHFFFAOYSA-N

Description

Synonym 1-Bromoisoquinolin-3-amine
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 144-146
Product Link https://store.apolloscientific.co.uk/product/3-amino-1-bromoisoquinoline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-1-bromoisoquinoline
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart