APO455831164 Apollo Scientific Ltd

1-Benzyl-4-chloropiperidine

  • MDL number MFCD00510091
  • Cas Number : 67848-71-9
  • Molecular Weight : 209.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-benzyl-4-chloropiperidine
Inchi Code 1S/C12H16ClN/c13-12-6-8-14(9-7-12)10-11-4-2-1-3-5-11/h1-5,12H,6-10H2
Inchi Key HEDDTLVBWCOBRG-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Liquid-Viscous
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-benzyl-4-chloropiperidine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-benzyl-4-chloropiperidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart