APO455831186 Apollo Scientific Ltd

2-Aminopyrimidine-4-carboxylic acid

  • MDL number MFCD07783936
  • Cas Number : 2164-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Amino-4-carboxypyrimidine, 2-Amino-4-carboxy-1,3-diazine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-aminopyrimidine-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-aminopyrimidine-4-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart