APO455831193 Apollo Scientific Ltd

(3S)-(+)-3-Hydroxytetrahydrofuran

  • MDL number MFCD00064327
  • Cas Number : 86087-23-2
  • Molecular Weight : 88.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 79
Iupac Name (3S)-tetrahydro-3-furanol
density 1.1
Inchi Code 1S/C4H8O2/c5-4-1-2-6-3-4/h4-5H,1-3H2/t4-/m0/s1
Inchi Key XDPCNPCKDGQBAN-BYPYZUCNSA-N

Description

Melting Point 79
Synonym (2S)-(+)-1,4-Epoxy-2-hydroxybutane, (3S)-(+)-Tetrahydrofuran-3-ol, (3S)-(+)-Oxolan-3-ol
Purity 99%
Density 1.1
Flash Point 80/15mm
Storage temp. Ambient Temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3s-3-hydroxytetrahydrofuran

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3s-3-hydroxytetrahydrofuran
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart