APO455831197 Apollo Scientific Ltd

4-Chloronicotinaldehyde

  • MDL number MFCD04112523
  • Cas Number : 114077-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Chloropyridine-3-carboxaldehyde, 4-Chloro-3-formylpyridine
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-chloronicotinaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloronicotinaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart