APO455831198 Apollo Scientific Ltd

4-Chloro-3-iodoaniline

  • MDL number MFCD11521292
  • Cas Number : 573764-31-5
  • Molecular Weight : 253.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-70
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloro-3-iodoaniline

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-3-iodoaniline
pictograms GHS06
hstatements H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart