APO455831202 Apollo Scientific Ltd

3-Bromo-5-iodophenol

  • MDL number MFCD12405397
  • Cas Number : 570391-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82-84
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=61432

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=61432
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart