APO455831204 Apollo Scientific Ltd

4-Chloro-5H-pyrrolo[3,2-d]pyrimidine

  • MDL number MFCD06658411
  • Cas Number : 84905-80-6
  • Molecular Weight : 153.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloro-5h-pyrrolo32-dpyrimidine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-5h-pyrrolo32-dpyrimidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart