APO455831207 Apollo Scientific Ltd

3-Bromo-5-nitrophenol

  • MDL number MFCD09965960
  • Cas Number : 116632-23-6
  • Molecular Weight : 218.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-5-nitrophenol
Inchi Code 1S/C6H4BrNO3/c7-4-1-5(8(10)11)3-6(9)2-4/h1-3,9H
Inchi Key VJQGLUHOAIZTNK-UHFFFAOYSA-N

Description

Synonym 3-Bromo-5-hydroxynitrobenzene
Purity 96%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-nitrophenol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-nitrophenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart