APO455831208 Apollo Scientific Ltd

3,6-Dimethyl-2-hydroxybenzaldehyde

  • MDL number MFCD04037328
  • Cas Number : 1666-04-2
  • Molecular Weight : 150.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3,6-Dimethyl-2-formylphenol, 2-Formyl-3-hydroxy-p-xylene, 3,6-Dimethylsalicylaldehyde
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/36-dimethyl-2-hydroxybenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/36-dimethyl-2-hydroxybenzaldehyde
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart