APO455831214 Apollo Scientific Ltd

4-Bromo-4'-(tert-butyl)biphenyl

  • MDL number MFCD01321141
  • Cas Number : 162258-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-143
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 152/2mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=61442

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=61442
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart