APO455831216 Apollo Scientific Ltd

4-Chloro-3-methoxybenzeneboronic acid

  • MDL number MFCD01318965
  • Cas Number : 89694-47-3
  • Molecular Weight : 186.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 232-234
Iupac Name 4-chloro-3-methoxyphenylboronic acid
Inchi Code 1S/C7H8BClO3/c1-12-7-4-5(8(10)11)2-3-6(7)9/h2-4,10-11H,1H3
Inchi Key DZNNRXURZJLARZ-UHFFFAOYSA-N

Description

Synonym (4-Chloro-3-methoxyphenyl)boronic acid
Purity 98%
Storage temp. Store at 2-8
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 232-234
Product Link https://store.apolloscientific.co.uk/product/4-chloro-3-methoxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-3-methoxybenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart