APO455831226 Apollo Scientific Ltd

5-Bromo-2-iodoanisole

  • MDL number MFCD11847482
  • Cas Number : 791642-68-7
  • Molecular Weight : 312.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-1-iodo-2-methoxybenzene, 5-Bromo-2-iodophenyl methyl ether
Purity 97
Physical Form Solid-Low Melt
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-iodoanisole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-iodoanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart