APO455831236 Apollo Scientific Ltd

3-Bromo-5-iodoaniline

  • MDL number MFCD09743665
  • Cas Number : 31948-87-5
  • Molecular Weight : 297.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 75-76
Iupac Name 3-bromo-5-iodoaniline
Inchi Code 1S/C6H5BrIN/c7-4-1-5(8)3-6(9)2-4/h1-3H,9H2
Inchi Key SCXCUTCIFLAPMW-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 75-76
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-iodoaniline

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-iodoaniline
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart