APO455831248 Apollo Scientific Ltd

Piperidine-4-sulphonyl chloride, N-CBZ protected

  • MDL number MFCD05664659
  • Cas Number : 287953-54-2
  • Molecular Weight : 317.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzyl 4-(chlorosulfonyl)-1-piperidinecarboxylate
Inchi Code 1S/C13H16ClNO4S/c14-20(17,18)12-6-8-15(9-7-12)13(16)19-10-11-4-2-1-3-5-11/h1-5,12H,6-10H2
Inchi Key HEGCWAOVTPVFBJ-UHFFFAOYSA-N

Description

Synonym N-[(Benzyloxy)carbonyl]piperidine-4-sulphonyl chloride, Benzyl 4-(chlorosulphonyl)piperidine-1-carboxylate
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/piperidine-4-sulphonyl-chloride-n-cbz-protected

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/piperidine-4-sulphonyl-chloride-n-cbz-protected
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart