APO455831250 Apollo Scientific Ltd

3-Bromoisoxazole

  • MDL number MFCD07778379
  • Cas Number : 111454-71-8
  • Molecular Weight : 147.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 60
Iupac Name 3-bromoisoxazole
density 1.812
Inchi Code 1S/C3H2BrNO/c4-3-1-2-6-5-3/h1-2H
Inchi Key GQMPHUXGZCDVGQ-UHFFFAOYSA-N

Description

Melting Point 60
Synonym 3-Bromo-1,2-oxazole
Density 1.812
Flash Point 182-183
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromoisoxazole

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromoisoxazole
pictograms GHS02;GHS07
hstatements H226;H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart