APO455831251 Apollo Scientific Ltd

4-Amino-3-methoxypyridine

  • MDL number MFCD09991938
  • Cas Number : 52334-90-4
  • Molecular Weight : 124.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-methoxy-4-pyridinamine
Inchi Code 1S/C6H8N2O/c1-9-6-4-8-3-2-5(6)7/h2-4H,1H3,(H2,7,8)
Inchi Key GPMHHELOMRCBRN-UHFFFAOYSA-N

Description

Synonym 3-Methoxypyridin-4-amine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-3-methoxypyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-3-methoxypyridine
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart