APO455831253 Apollo Scientific Ltd

4-Oxo-4H-pyran-2,6-dicarboxylic acid monohydrate

  • MDL number MFCD00149433
  • Cas Number : 6003-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 265
Synonym Chelidonic acid monohydrate, 2,6-Dicarboxy-4-oxo-4H-pyran monohydrate
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=49886

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=49886
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart