APO455831255 Apollo Scientific Ltd

1,5-Dibromo-2,4-dimethylbenzene

  • MDL number MFCD00154984
  • Cas Number : 615-87-2
  • Molecular Weight : 263.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71-72
Synonym 4,6-Dibromo-m-xylene
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/15-dibromo-24-dimethylbenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/15-dibromo-24-dimethylbenzene
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart