APO455831261 Apollo Scientific Ltd

Methyl 2-chloro-6-methylnicotinate

  • MDL number MFCD11845763
  • Cas Number : 53277-47-7
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-chloro-6-methylnicotinate
Inchi Code 1S/C8H8ClNO2/c1-5-3-4-6(7(9)10-5)8(11)12-2/h3-4H,1-2H3
Inchi Key SHKDOGHJQHIIRD-UHFFFAOYSA-N

Description

Synonym Methyl 2-chloro-6-methylpyridine-3-carboxylate, 2-Chloro-3-(methoxycarbonyl)-6-methylpyridine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-chloro-6-methylnicotinate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-chloro-6-methylnicotinate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart