APO455831274 Apollo Scientific Ltd

5-Bromo-2-iodobenzyl alcohol

  • MDL number MFCD09745850
  • Cas Number : 199786-58-8
  • Molecular Weight : 312.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-bromo-2-iodophenyl)methanol
Inchi Code 1S/C7H6BrIO/c8-6-1-2-7(9)5(3-6)4-10/h1-3,10H,4H2
Inchi Key LBMPOOHWJBYWPY-UHFFFAOYSA-N

Description

Synonym (5-Bromo-2-iodophenyl)methanol
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-iodobenzyl-alcohol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-iodobenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart