APO455831278 Apollo Scientific Ltd

2-Methyl-1-[4-(methylsulphanyl)phenyl]propan-1-one

  • MDL number MFCD03094661
  • Cas Number : 53207-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 32-33
Synonym 4-(Isobutanoyl)thioanisole, 4-(Isobutyryl)thioanisole, 4-(2-Methylpropanoyl)thioanisole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 122-124/2mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14981

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14981
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart