APO455831288 Apollo Scientific Ltd

2,6-Dimethoxyaniline

  • MDL number MFCD00053934
  • Cas Number : 2734-70-5
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 76-78
Iupac Name 2,6-dimethoxyaniline
Inchi Code 1S/C8H11NO2/c1-10-6-4-3-5-7(11-2)8(6)9/h3-5H,9H2,1-2H3
Inchi Key HQBJSEKQNRSDAZ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 76-78
Product Link https://store.apolloscientific.co.uk/product/26-dimethoxyaniline

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-dimethoxyaniline
pictograms GHS07;GHS08
hstatements H302+H332;H311;H373
Availability details for
Quantity
  • Availability
  • Add to Cart