APO455831293 Apollo Scientific Ltd

Methyl 3,5-dibromo-4-methylbenzoate

  • MDL number MFCD00010729
  • Cas Number : 74896-66-5
  • Molecular Weight : 307.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-90
Synonym Methyl 3,5-dibromo-p-toluate, 2,6-Dibromo-4-(methoxycarbonyl)toluene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-35-dibromo-4-methylbenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-35-dibromo-4-methylbenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart