APO455831296 Apollo Scientific Ltd

4-Bromobenzhydrol

  • MDL number MFCD00016851
  • Cas Number : 29334-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61-63
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromobenzhydrol

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS https://store.apolloscientific.co.uk/product/4-bromobenzhydrol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart