APO455831298 Apollo Scientific Ltd

2'-Methylacetanilide

  • MDL number MFCD00014961
  • Cas Number : 120-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109-110
Synonym N-(2-Methylphenyl)acetamide, 2-Acetamidotoluene
Purity 98
Density 1.16
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 296
Product Link https://store.apolloscientific.co.uk/product/2-methylacetanilide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-methylacetanilide
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart