APO455831300 Apollo Scientific Ltd

Methyl 4-iodo-3-methylbenzoate

  • MDL number MFCD02683863
  • Cas Number : 5471-81-8
  • Molecular Weight : 276.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-62
Synonym 2-Iodo-5-(methoxycarbonyl)toluene, Methyl 4-iodo-m-toluate
Physical Form Solid-Crystals
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-4-iodo-3-methylbenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-iodo-3-methylbenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart