APO455831316 Apollo Scientific Ltd

3-Bromopyridine-2,6-diamine

  • MDL number MFCD04113819
  • Cas Number : 54903-86-5
  • Molecular Weight : 188.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171-176
Synonym 3-Bromo-2,6-diaminopyridine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromopyridine-26-diamine

Safety Information

signalword Warning
pcodes P280;P301+P312;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/3-bromopyridine-26-diamine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart