APO455831321 Apollo Scientific Ltd

3-Amino-6-chloropyridazine

  • MDL number MFCD00051506
  • Cas Number : 5469-69-2
  • Molecular Weight : 129.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 226-228
Synonym 6-Chloropyridazin-3-amine, 3-Amino-6-chloro-1,2-diazine
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-6-chloropyridazine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-6-chloropyridazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart