APO455831327 Apollo Scientific Ltd

2-(Dimethoxymethyl)-1,6-naphthyridine

  • MDL number MFCD00202920
  • Cas Number : 386715-36-2
  • Molecular Weight : 204.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,6-Naphthyridine-2-carboxaldehyde dimethyl acetal
Purity 95
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-dimethoxymethyl-16-naphthyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-dimethoxymethyl-16-naphthyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart