APO455831329 Apollo Scientific Ltd

5-Bromo-2-furohydrazide

  • MDL number MFCD01310787
  • Cas Number : 89282-37-1
  • Molecular Weight : 205.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 136-138
Iupac Name 5-bromo-2-furohydrazide
Inchi Code 1S/C5H5BrN2O2/c6-4-2-1-3(10-4)5(9)8-7/h1-2H,7H2,(H,8,9)
Inchi Key AKANWSCBFOMJNS-UHFFFAOYSA-N

Description

Synonym 5-Bromo-2-furoic acid hydrazide, 5-Bromofuran-2-carbohydrazide
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 136-138
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-furohydrazide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-furohydrazide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart