APO455831331 Apollo Scientific Ltd

2-(Methylsulphonyl)aniline hydrochloride

  • MDL number MFCD00216484
  • Cas Number : 205985-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-141
Synonym 2-Aminophenyl methyl sulphone hydrochloride
Purity 95
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15342

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=15342
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart