APO455831336 Apollo Scientific Ltd

5-Aminobenzo[b]thiophene

  • MDL number MFCD04115381
  • Cas Number : 20532-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72-75
Synonym 1-Benzothiophen-5-amine, 5-Amino-1-benzothiophene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-aminobenzobthiophene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-aminobenzobthiophene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart