APO455831342 Apollo Scientific Ltd

Methyl 3-bromo-5-chloropyridine-2-carboxylate

  • MDL number MFCD14698105
  • Cas Number : 1214362-45-4
  • Molecular Weight : 250.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-bromo-5-chloro-2-pyridinecarboxylate
Inchi Code 1S/C7H5BrClNO2/c1-12-7(11)6-5(8)2-4(9)3-10-6/h2-3H,1H3
Inchi Key FGGMRJLIUFDXBD-UHFFFAOYSA-N

Description

Synonym 3-Bromo-5-chloro-2-(methoxycarbonyl)pyridine, Methyl 3-bromo-5-chloropicolinate
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-bromo-5-chloropyridine-2-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-bromo-5-chloropyridine-2-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart