APO455831354 Apollo Scientific Ltd

Isoquinoline-6-carbonitrile

  • MDL number MFCD09991660
  • Cas Number : 106778-42-1
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-isoquinolinecarbonitrile
Inchi Code 1S/C10H6N2/c11-6-8-1-2-10-7-12-4-3-9(10)5-8/h1-5,7H
Inchi Key DCSWGUBEMVRKQO-UHFFFAOYSA-N

Description

Synonym 6-Cyanoisoquinoline
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/isoquinoline-6-carbonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/isoquinoline-6-carbonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart