APO455831356 Apollo Scientific Ltd

4-Hydroxyisoquinoline

  • MDL number MFCD00234514
  • Cas Number : 3336-49-0
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-isoquinolinol
Inchi Code 1S/C9H7NO/c11-9-6-10-5-7-3-1-2-4-8(7)9/h1-6,11H
Inchi Key DXTUTXYCHFWRQC-UHFFFAOYSA-N

Description

Synonym Isoquinolin-4-ol, 4-Hydroxy-2-azanaphthalene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-hydroxyisoquinoline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxyisoquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart