APO455831358 Apollo Scientific Ltd

Isoquinoline-6-carboxylic acid

  • MDL number MFCD09910323
  • Cas Number : 106778-43-2
  • Molecular Weight : 173.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Carboxyisoquinoline, 6-Carboxy-2-azanaphthalene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/isoquinoline-6-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/isoquinoline-6-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart