APO455831364 Apollo Scientific Ltd

3-Amino-4-methylbenzonitrile

  • MDL number MFCD04039781
  • Cas Number : 60710-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-77
Synonym 5-Cyano-2-methylaniline, 2-Amino-4-cyanotoluene, 5-Cyano-o-toluidine
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-amino-4-methylbenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-4-methylbenzonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart