APO455831368 Apollo Scientific Ltd

N,N'-(Benzene-1,4-diyl)dibenzenesulphonamide

  • MDL number MFCD00183146
  • Cas Number : 16504-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N1,N4-Bis(phenylsulphonyl)benzene-1,4-diamine, N,N'-(1,4-Phenylene)dibenzenesulphonamide
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=51321

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=51321
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart