APO455831370 Apollo Scientific Ltd

5-Nitro-1H-indole-2-carboxylic acid

  • MDL number MFCD00236372
  • Cas Number : 16730-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >300
Synonym 2-Carboxy-5-nitro-1H-indole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-nitro-1h-indole-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-nitro-1h-indole-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart