APO455831379 Apollo Scientific Ltd

2',4',6'-Trihydroxyacetophenone

  • MDL number MFCD00002287
  • Cas Number : 480-66-0
  • Molecular Weight : 168.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 219-221
Iupac Name 1-(2,4,6-trihydroxyphenyl)ethanone
Inchi Code 1S/C8H8O4/c1-4(9)8-6(11)2-5(10)3-7(8)12/h2-3,10-12H,1H3
Inchi Key XLEYFDVVXLMULC-UHFFFAOYSA-N

Description

Synonym 1-(2,4,6-Trihydroxyphenyl)ethan-1-one, 2-Acetylbenzene-1,3,5-triol
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 219-221
Product Link https://store.apolloscientific.co.uk/product/246-trihydroxyacetophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/246-trihydroxyacetophenone
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart