APO455831389 Apollo Scientific Ltd

3-Nitro-1H-1,2,4-triazole

  • MDL number MFCD00009749
  • Cas Number : 24807-55-4
  • Molecular Weight : 114.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210(dec.)
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-nitro-1h-124-triazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-nitro-1h-124-triazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart