APO455831392 Apollo Scientific Ltd

6-Chloro-2-methylimidazo[1,2-a]pyridine-3-carboxylic acid

  • MDL number MFCD00269625
  • Cas Number : 138642-96-3
  • Molecular Weight : 210.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-2-methylimidazo[1,2-a]pyridine-3-carboxylic acid
Inchi Code 1S/C9H7ClN2O2/c1-5-8(9(13)14)12-4-6(10)2-3-7(12)11-5/h2-4H,1H3,(H,13,14)
Inchi Key PGYYUAYGIWHUJA-UHFFFAOYSA-N

Description

Synonym 3-Carboxy-6-chloro-2-methylimidazo[1,2-a]pyridine
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-chloro-2-methylimidazo12-apyridine-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/6-chloro-2-methylimidazo12-apyridine-3-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart