APO455831402 Apollo Scientific Ltd

6-Hydroxynicotinaldehyde

  • MDL number MFCD03094941
  • Cas Number : 106984-91-2
  • Molecular Weight : 123.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 222-226
Iupac Name 6-oxo-5,6-dihydro-3-pyridinecarbaldehyde
Inchi Code 1S/C6H5NO2/c8-4-5-1-2-6(9)7-3-5/h1-4H,(H,7,9)
Inchi Key BUMAFTGGYPBHHK-UHFFFAOYSA-N

Description

Synonym 6-Hydroxypyridine-3-carboxaldehyde, 5-Formyl-2-hydroxypyridine
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 222-226
Product Link https://store.apolloscientific.co.uk/product/6-hydroxynicotinaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-hydroxynicotinaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart