APO455831404 Apollo Scientific Ltd

3-(tert-Butyl)aniline

  • MDL number MFCD00125078
  • Cas Number : 5369-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Amino-3-(tert-butyl)benzene
Purity 98
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 140/30mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13506

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13506
pictograms GHS07
hstatements H302;H315;H319;H332
Availability details for
Quantity
  • Availability
  • Add to Cart