APO455831408 Apollo Scientific Ltd

4-(4-Chlorophenyl)pyrimidin-2-amine

  • MDL number MFCD00665913
  • Cas Number : 133256-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-174
Synonym 2-Amino-4-(4-chlorophenyl)pyrimidine, 2-Amino-4-(4-chlorophenyl)-1,3-diazine
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=12571

Safety Information

signalword Warning
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=12571
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart