APO455831415 Apollo Scientific Ltd

5-(Isoxazol-5-yl)thiophene-2-sulphonyl chloride

  • MDL number MFCD02187482
  • Cas Number : 551930-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69-70
Synonym 5-(1,2-Oxazol-5-yl)thiophene-2-sulphonyl chloride, 2-(Chlorosulphonyl)-5-(isoxazol-5-yl)thiophene, 5-[5-(Chlorosulphonyl)thien-2-yl]-1,2-oxazole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15014

Safety Information

signalword Danger
pcodes P280;P260
MSDS http://www.apolloscientific.co.uk/display_item.php?id=15014
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart