APO455831432 Apollo Scientific Ltd

4-(tert-Butyl)-2-nitroaniline

  • MDL number MFCD00066305
  • Cas Number : 6310-19-6
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 101-105
Iupac Name 4-tert-butyl-2-nitroaniline
Inchi Code 1S/C10H14N2O2/c1-10(2,3)7-4-5-8(11)9(6-7)12(13)14/h4-6H,11H2,1-3H3
Inchi Key YXTJXBPEGJNGKV-UHFFFAOYSA-N

Description

Synonym 2-Amino-5-(tert-butyl)nitrobenzene
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 101-105
Product Link https://store.apolloscientific.co.uk/product/4-tert-butyl-2-nitroaniline

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-tert-butyl-2-nitroaniline
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart